missing translation for 'onlineSavingsMsg'
Learn More

Ergodyne™ N-Ferno™ 6471 Thermal Bibs/Overalls

Catalog No. 19910199
Change view
Click to view available options
Size:
Small
Medium
Large
X-Large
2X-Large
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
19910199 -
19910194 Small
19910195 Medium
19910196 Large
19910197 X-Large
19910198 2X-Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Catalog No. 19910199 Supplier Ergodyne™ Supplier No. 41217

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Insulated bibs/overalls provide the ultimate in durability, weather protection, and quality outerwear

Features removable knee/shin pads and 1600D nylon reinforcement in heavy wear areas

Specifications

Color Black
Material Nylon (500D)
For Use With (Application) Roof Bolting, Mining, Oil/Gas, Demolition, Heavy Construction, Ironwork, Mechanics