Learn More
Fisherbrand™ Multistage Gas Cylinder Regulators
For Noncorrosive General Purpose Gases. Diaphragm-Sensed Dual-Stage Regulators designed for consistent outlet pressure delivery.
$466.58 - $849.88
Specifications
Bonnet Material Brass
Connection Type Outlet needle valve with 0.25 in. NPTM thread
Diaphragm Material Neoprene
Type Cylinder Regulator
For Use With (Application) Noncorrosive gases, requiring maintenance of a constant delivery pressure until cylinder contents are empty
View More Specs
Alert:
Not for use with corrosive, toxic or high-purity gases.
Products 21
Catalog Number Mfr. No. Inlet Delivery Pressure Price Quantity & Availability  
Catalog Number Mfr. No. Inlet Delivery Pressure Price Quantity & Availability  
10575142
Accessories View Documents Promotion Details
Fisherbrand™
10575142
CGA 350 1 to 125 psi
Each for $678.85
Only null left
10575133
View Documents Promotion Details
Fisherbrand™
10575133
CGA 346 1 to 40 psi
Each for $466.58
Only null left
10575150
Accessories View Documents Promotion Details
Fisherbrand™
10575150
CGA 500 1 to 125 psi
Each for $678.85
Only null left
10575136
Accessories View Documents Promotion Details
Fisherbrand™
10575136
CGA 580 1 to 125 psi
Each for $707.85
Only null left
10575152
View Documents Promotion Details
Fisherbrand™
10575152
CGA 296 1 to 125 psi
Each for $702.03
Only null left
10575139
Accessories View Documents Promotion Details
Fisherbrand™
10575139
CGA 320 1 to 40 psi
Each for $729.30
Only null left
Edge
10575147
Accessories View Documents Promotion Details
Fisherbrand™
10575147
CGA 280 1 to 40 psi
Each for $668.06
Only null left
10575140
Accessories View Documents Promotion Details
Fisherbrand™
10575140
CGA 320 1 to 125 psi
Each for $816.68
Only null left
10575151
View Documents Promotion Details
Fisherbrand™
10575151
CGA 296 1 to 40 psi
Each for $713.15
Only null left
10575143
View Documents Promotion Details
Fisherbrand™
10575143
CGA 326 1 to 40 psi
Each for $599.76
Only null left
10575149
View Documents Promotion Details
Fisherbrand™
10575149
CGA 500 1 to 40 psi
Each for $634.44
Only null left
10575144
Accessories View Documents Promotion Details
Fisherbrand™
10575144
CGA 540 1 to 40 psi
Each for $734.53
Only null left
10575146
Accessories View Documents Promotion Details
Fisherbrand™
10575146
CGA 540 1 to 325 psi
Each for $727.32
Only null left
10575131
Accessories View Documents Promotion Details
Fisherbrand™
10575131
CGA 510 1 to 15 psi
Each for $706.26
Only null left
10575141
View Documents Promotion Details
Fisherbrand™
10575141
CGA 350 1 to 40 psi
Each for $849.88
Only null left
10575134
Accessories View Documents Promotion Details
Fisherbrand™
10575134
CGA 590 1 to 40 psi
Each for $727.32
Only null left
10575148
View Documents Promotion Details
Fisherbrand™
10575148
CGA 280 1 to 125 psi
Each for $678.85
Only null left
10575137
Accessories View Documents Promotion Details
Fisherbrand™
10575137
CGA 580 1 to 250 psi
Each for $734.79
Only null left
10575138
View Documents Promotion Details
Fisherbrand™
10575138
CGA 580 1 to 325 psi
Each for $734.79
Only null left
10575145
Accessories View Documents Promotion Details
Fisherbrand™
10575145
CGA 540 1 to 125 psi
Each for $680.24
Only null left
10575135
Accessories View Documents Promotion Details
Fisherbrand™
10575135
CGA 580 1 to 40 psi
Each for $721.74
Only null left
Description
Description
For Noncorrosive General Purpose Gases. Diaphragm-Sensed Dual-Stage Regulators designed for consistent outlet pressure delivery.
Ideal for applications requiring maintenance of a constant delivery pressure until cylinder contents are close to empty.
  • Feature Neoprene diaphragms reinforced with nylon fabric, nylon seals, Orthane™ seats for first and second stages
  • Dual-scale gauges (PSI/KPa), brass bonnet, External self-reseating relief valve and easy pressure-adjusting knob
  • Delrin Cap Bushing for smooth pressure adjustment
  • Outlet Needle Valve with 1/4 in. NPT male thread
Specifications
Specifications
Brass
Neoprene
Noncorrosive gases, requiring maintenance of a constant delivery pressure until cylinder contents are empty
Multi
Nylon Fabric, Nylon
Outlet needle valve with 0.25 in. NPTM thread
Cylinder Regulator
Dual-scale
Yes
Orthane™