missing translation for 'onlineSavingsMsg'
Learn More

Fisherbrand™ Multistage Gas Cylinder Regulators

For Noncorrosive General Purpose Gases. Diaphragm-Sensed Dual-Stage Regulators designed for consistent outlet pressure delivery.

Supplier:  Fisherbrand™ 10575144

 View more versions of this product

Catalog No. 10-575-144


Explore available promotions

Alert:

Not for use with corrosive, toxic or high-purity gases.


Description

Description

For Noncorrosive General Purpose Gases. Diaphragm-Sensed Dual-Stage Regulators designed for consistent outlet pressure delivery.

Ideal for applications requiring maintenance of a constant delivery pressure until cylinder contents are close to empty.

  • Feature Neoprene diaphragms reinforced with nylon fabric, nylon seals, Orthane™ seats for first and second stages
  • Dual-scale gauges (PSI/KPa), brass bonnet, External self-reseating relief valve and easy pressure-adjusting knob
  • Delrin Cap Bushing for smooth pressure adjustment
  • Outlet Needle Valve with 1/4 in. NPT male thread
Specifications

Specifications

Brass
1 to 40 psi
Cylinder Regulator
Oxygen
540
2 to 60 psi
Yes
Orthane™
Outlet needle valve with 0.25 in. NPTM thread
Neoprene
Noncorrosive gases, requiring maintenance of a constant delivery pressure until cylinder contents are empty
Dual-scale
Multi
3000 psi
Nylon Fabric, Nylon
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Accessories
Promotions

Promotions