missing translation for 'onlineSavingsMsg'
Learn More

Fisherbrand™ Multistage Gas Cylinder Regulators

Catalog No. 10575131
Change view
Click to view available options
Inlet:
CGA 280
CGA 296
CGA 320
CGA 326
CGA 346
CGA 350
CGA 500
CGA 510
CGA 540
CGA 580
CGA 590
Delivery Pressure:
1 to 125 psi
1 to 15 psi
1 to 250 psi
1 to 325 psi
1 to 40 psi
21 product options available for selection
Product selection table with 21 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Inlet Delivery Pressure
10575131 CGA 510 1 to 15 psi
10575142 CGA 350 1 to 125 psi
10575133 CGA 346 1 to 40 psi
10575150 CGA 500 1 to 125 psi
10575136 CGA 580 1 to 125 psi
10575152 CGA 296 1 to 125 psi
10575139 CGA 320 1 to 40 psi
10575147 CGA 280 1 to 40 psi
10575140 CGA 320 1 to 125 psi
10575151 CGA 296 1 to 40 psi
10575143 CGA 326 1 to 40 psi
10575149 CGA 500 1 to 40 psi
10575144 CGA 540 1 to 40 psi
10575146 CGA 540 1 to 325 psi
10575141 CGA 350 1 to 40 psi
10575134 CGA 590 1 to 40 psi
10575148 CGA 280 1 to 125 psi
10575137 CGA 580 1 to 250 psi
10575138 CGA 580 1 to 325 psi
10575145 CGA 540 1 to 125 psi
10575135 CGA 580 1 to 40 psi
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 21 options available.
21 options

Not for use with corrosive, toxic or high-purity gases.

Catalog No. 10575131 Supplier Fisherbrand™ Supplier No. 10575131

Not for use with corrosive, toxic or high-purity gases.

For Noncorrosive General Purpose Gases. Diaphragm-Sensed Dual-Stage Regulators designed for consistent outlet pressure delivery.

For Noncorrosive General Purpose Gases. Diaphragm-Sensed Dual-Stage Regulators designed for consistent outlet pressure delivery.

Ideal for applications requiring maintenance of a constant delivery pressure until cylinder contents are close to empty.

  • Feature Neoprene diaphragms reinforced with nylon fabric, nylon seals, Orthane™ seats for first and second stages
  • Dual-scale gauges (PSI/KPa), brass bonnet, External self-reseating relief valve and easy pressure-adjusting knob
  • Delrin Cap Bushing for smooth pressure adjustment
  • Outlet Needle Valve with 1/4 in. NPT male thread

Specifications

Bonnet Material Brass
Connection Type Outlet needle valve with 0.25 in. NPTM thread
Delivery Pressure 1 to 15 psi
Diaphragm Material Neoprene
Type Cylinder Regulator
For Use With (Application) Noncorrosive gases, requiring maintenance of a constant delivery pressure until cylinder contents are empty
Gases Measured Acetylene
Gauge Dual-scale
Inlet CGA 510
No. of Stages Multi
Outlet Gauge Pressure 2 to 30 psi
Pressure Range (Inlet) 400 psi
Relief Valve Yes
Seal Material Nylon Fabric, Nylon
Seat Material Orthane™
Show More Show Less