missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MilliporeSigma™ MQuant™ Total Hardness Test Strips
To determine the total hardness of water
Supplier: MilliporeSigma™ 1.10047.0013
Comprend: Tube containing 100 strips
Description
- Small and easy to handle
- Flexible and universal application
- EDTA reaction method ideal for screening
- Immediate result produced through on-site analysis
- PET film and low reagent content
- Easily disposable
1.10025.0001
Color-scale graduation: 4 - 5 - 9 - 18 - 26°e
1.10046.0001
Color-scale graduation: 6 - 13 - 19 - 25 - 31°e
Reclose the tube containing the test strips immediately after use.
Analytical Quality Assurance, Standard for Total Hardness
Identifiants chimiques
| Analytical quality assurance, to determine the total hardness of water | |
| 15°C to 25°C | |
| Test Strip |
Spécifications
| Analytical quality assurance, to determine the total hardness of water | |
| 6 to 31°e | |
| 6 to 31°e | |
| EDTA |
| Colorimeter | |
| 25000 | |
| 15°C to 25°C | |
| Test Strip |