missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MilliporeSigma™ MQuant™ Total Hardness Test Strips
Description
Includes
Tube containing 100 strips
- Small and easy to handle
- Flexible and universal application
- EDTA reaction method ideal for screening
- Immediate result produced through on-site analysis
- PET film and low reagent content
- Easily disposable
1.10025.0001
Color-scale graduation: 4 - 5 - 9 - 18 - 26°e
1.10046.0001
Color-scale graduation: 6 - 13 - 19 - 25 - 31°e
Reclose the tube containing the test strips immediately after use.
Analytical Quality Assurance, Standard for Total Hardness
Specifications
Specifications
| For Use With (Application) | Analytical quality assurance, to determine the total hardness of water |
| For Use With (Equipment) | Colorimeter |
| Graduations | 6 to 31°e |
| No. of Tests | 100 |
| Range | 6 to 31°e |
| Storage Requirements | 15°C to 25°C |
| Test Method | EDTA |
| Type | Test Strip |