missing translation for 'onlineSavingsMsg'
Learn More

MilliporeSigma mPAGE™ Lux Casting System

Casting System for hand casting mini gels using UV light with a 0.75 mm, 1.0 mm or 1.5 mm thickness, includes instrument, casters, plates and combs

$2,780.00

Specifications

For Use With (Application) Hand Casting Mini Gels
Gel Size 8.3 x 10.1 cm
Max. Current 1.5 A
Type Gel Casting System
Max. Voltage 240 VAC
View More Specs

Products 3
Catalog Number Mfr. No. Thickness (Metric) Price Quantity & Availability  
Catalog Number Mfr. No. Thickness (Metric) Price Quantity & Availability  
11580610
View Documents Promotion Details
MilliporeSigma
LUXCSYS15MUS
1.5 mm
Pack for $2,780.00
Only null left
 
11580611
View Documents Promotion Details
MilliporeSigma
LUXCSYS1MUS
1.0 mm
Pack for $2,780.00
Only null left
 
11580612
View Documents Promotion Details
MilliporeSigma
LUXCSYS75MUS
0.75 mm
Pack for $2,780.00
Only null left
 
Description

Description

  • The mPAGE™ Lux Casting System offers rapid and simple gel curing process for handcasting mini protein gels
  • A mini gel is cured in 1.5 min and overall casting process can be less than 3 min
  • Gels cast are 8 cm x 10 cm with a gel thickness of 1.0 mm (40 μL), 0.75 mm (25 μL), or 1.5 mm (60 μL) for 10-well option sample loading
  • Gels cast are 8 cm x 10 cm with a gel thickness of 1.0 mm (25 μL) or 1.5 mm (36 μL), for 15-well option sample loading
Specifications

Specifications

Hand Casting Mini Gels
1.5 A
240 VAC
8.3 x 10.1 cm
Gel Casting System
Videos
SDS
Documents

Documents