Learn More
MilliporeSigma mPAGE™ Lux Casting System
Casting System for hand casting mini gels using UV light with a 0.75 mm, 1.0 mm or 1.5 mm thickness, includes instrument, casters, plates and combs
Supplier: MilliporeSigma LUXCSYS1MUS
Description
- The mPAGE™ Lux Casting System offers rapid and simple gel curing process for handcasting mini protein gels
- A mini gel is cured in 1.5 min and overall casting process can be less than 3 min
- Gels cast are 8 cm x 10 cm with a gel thickness of 1.0 mm (40 μL), 0.75 mm (25 μL), or 1.5 mm (60 μL) for 10-well option sample loading
- Gels cast are 8 cm x 10 cm with a gel thickness of 1.0 mm (25 μL) or 1.5 mm (36 μL), for 15-well option sample loading
Chemical Identifiers
| Hand Casting Mini Gels | |
| 1.5 A | |
| 240 VAC |
| 8.3 x 10.1 cm | |
| Gel Casting System |
Specifications
| Hand Casting Mini Gels | |
| 8.3 x 10.1 cm | |
| Gel Casting System | |
| 240 VAC |
| Lux Bis-Tris Reagent Kitm Lux Masked Short Plates Lux Mixing Tubes Combs Mini Spacer Plates Mini Gel Tank Mini Wet Transfer System | |
| 1.5 A | |
| 1.0 mm |