missing translation for 'onlineSavingsMsg'
Learn More

MilliporeSigma mPAGE™ Lux Casting System

Catalog No. 11580610
Change view
Click to view available options
Thickness (Metric):
0.75 mm
1.0 mm
1.5 mm
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Thickness (Metric)
11580610 1.5 mm
11580611 1.0 mm
11580612 0.75 mm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 11580610 Supplier MilliporeSigma Supplier No. LUXCSYS15MUS

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Casting System for hand casting mini gels using UV light with a 0.75 mm, 1.0 mm or 1.5 mm thickness, includes instrument, casters, plates and combs

  • The mPAGE™ Lux Casting System offers rapid and simple gel curing process for handcasting mini protein gels
  • A mini gel is cured in 1.5 min and overall casting process can be less than 3 min
  • Gels cast are 8 cm x 10 cm with a gel thickness of 1.0 mm (40 μL), 0.75 mm (25 μL), or 1.5 mm (60 μL) for 10-well option sample loading
  • Gels cast are 8 cm x 10 cm with a gel thickness of 1.0 mm (25 μL) or 1.5 mm (36 μL), for 15-well option sample loading

Specifications

For Use With (Application) Hand Casting Mini Gels
For Use With (Equipment) Lux Bis-Tris Reagent Kit Lux Masked Short Plates Lux Mixing Tubes Combs Mini Spacer Plates Mini Gel Tank Mini Wet Transfer System
Gel Size 8.3 x 10.1 cm
Max. Current 1.5 A
Type Gel Casting System
Thickness (Metric) 1.5 mm
Max. Voltage 240 VAC