missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MP Biomedicals™ Hyaluronidase, Bovine Testes
Description
Hyaluronidase, [37326-33-3]
- Hyaluronate Glycanohydrolase
- Activity: 300 USP units/mg min.
- Lyophilized powder
Specifications
Specifications
| Concentration | ≥300 U/mg |
| Color | Tan |
| pH | 4.5 to 6 |
| For Use With (Application) | Drug dispersion and delivery |
| Content And Storage | -20°C |
| Source | Bovine testes |
| Quantity | 100 mg |
| Product Type | Hyaluronidase |
| Molecular Weight (g/mol) | ≈55 |
| Inhibitors | Fe2+ and Fe3+ are inhibitory as are Mn2+ and Cu2+ |
| Show More |