missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Mouse RANTES ELISA Development Kit (ABTS), PeproTech®

Numéro de catalogue. 900K124K
Change view
Click to view available options
Quantity:
10 x 96 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Numéro de catalogue. Quantity
900K124K 10 x 96 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Numéro de catalogue. 900K124K Fournisseur Invitrogen™ Code fournisseur 900K124K
Il en reste null

ELISA

CCL5 (RANTES) is a chemokine gene located on the q-arm of chromosome 17. Chemokines are a superfamily of secreted proteins involved in immunoregulatory and inflammatory processes. The superfamily is divided into four subfamilies based on the arrangement of the N-terminal cysteine residues of the mature peptide. CCL5, a member of the CC subfamily, functions as a chemoattractant for blood monocytes, memory T helper cells, and eosinophils. It causes the release of histamine from basophils and activates eosinophils. This cytokine is one of the major HIV-suppressive factors produced by CD8+ cells. It functions as one of the natural ligands for the chemokine receptor chemokine (C-C motif) receptor 5 (CCR5), and it suppresses in vitro replication of the R5 strains of HIV-1, which use CCR5 as a coreceptor. Alternative splicing results in multiple transcript variants that encode different isoforms. Diseases associated with CCL5 include Human Immunodeficiency Virus Type 1 and Periapical Granuloma.

Spécifications

Accession Number P30882
Assay Range Lot-specific
Assay Sensitivity Lot-specific
Conjugate HRP
Product Type ELISA
For Use With (Equipment) Colorimetric Microplate Reader
Gene ID (Entrez) 20304
Gene Symbol CCL5
Kit Contents Capture Antibody, Detection Antibody, Protein Standard, HRP-conjugate
Label or Dye HRP
Quantity 10 x 96 Tests
Regulatory Status RUO
Storage Requirements Refer to product documentation for component specific storage temperature.
Target CCL5 (RANTES)
Test Time 1 hrs 45 mins
Total Assay Time 24 hrs 30 mins
Afficher plus de résultats Afficher moins de résultats