missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Mouse IL-17BR ELISA Kit

Numéro de catalogue. EM42RBX5
Encompass
Change view
Click to view available options
Quantity:
5 x 96 Tests
96 Tests
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Numéro de catalogue. Quantity
EM42RBX5 5 x 96 Tests
EM42RB 96 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Numéro de catalogue. EM42RBX5 Fournisseur Invitrogen™ Code fournisseur EM42RBX5
Il en reste null

ELISA

Mouse IL-17BR quantitates mouse IL-17BR in serum, plasma, supernatant. The assay will exclusively recognize both natural and recombinant mouse IL-17BR.

Recently, a number of cytokines belonging to the interleukin (IL)-17 family have been identified. These are termed as IL-17B, IL-17C (1) and IL-17E. IL-17 is a potent proinflammatory cytokine that plays roles in a number of diseases including rheumatoid arthritis, multiple sclerosis, and promotion of tumor growth. IL-17B, C, and E like IL-17 are able to induce proinflammatory responses. However, they do not bind to the IL-17 receptor suggesting that additional IL-17R related receptor might exist. Receptor for IL-17B and IL-17E has been independently isolated and has been designated as EV127 (in mouse) and IL-17Rh1 (in human), respectively. IL-17E induces activation of NF-kB pathway and like IL-17 also induces production of IL-8. IL-17Rh1 mRNA is highly expressed in Kidney and liver and at moderate level in small intestine, colon, and brain.

Spécifications

Accession Number CRL4; cytokine receptor CRL4; cytokine receptor-like 4; ecotropic viral integration site 27; Evi27; IL-17 receptor B; IL-17 receptor homolog 1; IL-17 B; IL-17 B receptor; IL17BR; IL-17 ER; IL17RB; IL-17 RB; IL17RH1; IL-17 Rh1; IL-20; interleukin 17 receptor B; interleukin 17 receptor homolog 1; interleukin 17 B receptor; Interleukin-17 receptor B; interleukin-17 B receptor; MGC138900; MGC138901; MGC5245; UNQ2501/PRO19612; ZCYTO7
Assay Range 0.2 to 50 ng/mL
Assay Sensitivity 0.2 ng/mL
Conjugate HRP
Product Type ELISA
Sample Type Plasma, Serum, Supernatant
For Use With (Equipment) Colorimetric Microplate Reader
Gene ID (Entrez) 50905
Gene Symbol IL17RB
Interassay CV <12%
Intraassay CV <10
Kit Contents Pre-coated 96 well plate, Standard, Assay Diluent concentrate, Biotinylated Detection Antibody, SAV-HRP, Wash Buffer, Chromogen, Stop Solution, Adhesive Plate Covers
Label or Dye HRP
Quantity 5 x 96 Tests
Regulatory Status RUO
Sample Volume Plasma 50 μL, Serum 50 μL, Supernatant 50 μL
Storage Requirements 2°C to 8°C
Target IL17RB
Test Time 1 hrs 20 mins
Total Assay Time 4 hrs 45 mins
Afficher plus de résultats Afficher moins de résultats