missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Mouse IL-17A (homodimer) Uncoated ELISA Kit with Plates

ELISA

Supplier:  Invitrogen™ 88737186

 Afficher plus de versions de ce produit

Numéro de catalogue. 501128984


Il en reste null

Description

Description

The Mouse Interleukin 17A Homodimer (IL-17A) Uncoated ELISA Kit contains pre-matched antibody pairs, plates and reagents for performing quantitative enzyme linked immunosorbent assays (ELISA) to detect and quantify protein levels of mouse IL-17A. Wash Buffer and Stop Solution are needed to complete the ELISA reaction and are sold separately. Principle of the method ELISAs are designed to measure the amount of the target bound between a matched antibody pair. A target-specific antibody is coated to the bottom of the wells of a microplate, which is an overnight process. Samples, standards, or controls are then added into these wells and bind to the immobilized (capture) antibody. A sandwich is formed by the addition of the second (detector) antibody, a substrate solution is added that reacts with the enzyme-antibody-target complex to produce measurable signal. The intensity of this signal is directly proportional to the concentration of target present in the original specimen.

Interleukin-17A (IL-17A, CTLA-8) is a CD4+ T cell-derived cytokine that promotes inflammatory responses in cell lines and is elevated in rheumatoid arthritis, asthma, multiple sclerosis, psoriasis, and transplant rejection. IL-17A is a 32 kDa long, disulfide-linked homodimer consisting of 136 amino acids that is a member of a six-species family of proteins (IL-17A-17F) and signals through the IL-17 receptor (IL-17R/CDw217). High levels of IL-17A homodimer are produced by activated peripheral blood CD4+ T-cells, and IL-17A also enhances expression of the intracellular adhesion molecule-1 (ICAM-1) in human fibroblasts. In particular, human IL-17A also stimulates epithelial, endothelial, or fibroblastic cells to secrete IL-6, IL-8, G-CSF, and PGE2. In the presence of human IL-17A, fibroblasts can sustain the proliferation of CD34+ hematopoietic progenitors and induce maturation into neutrophils. Mouse, rat, and human IL-17A can induce IL-6 secretion in mouse stromal cells, indicating that all homologs can recognize the mouse receptor. IL-17A regulates the activities of NF-kappa B and mitogen-activated protein kinases, stimulates the expression of IL-6 and cyclooxygenase-2 (PTGS2/COX-2), and enhances the production of nitric oxide (NO).
Spécifications

Identifiants chimiques

Q62386
8 pg/mL
ELISA
Mouse
Absorbance Microplate Reader
IL17A
Biotin
Serum 100 μL, Supernatant 100 μL
Interleukin-17 A, IL-17 AA
1 hr 45 min
8-500 pg/mL
HRP
Serum, Supernatant
ELISA
16171
Capture Antibody: Pre-titrated, purified antibody, Detection Antibody: Pre-titrated, HRP-conjugated antibody, Standard: Recombinant protein for generating standard curve and calibrating samples, 10X Coating Buffer: Buffer for plating the Capture Antibody, 5X ELISA/ELISPOT Diluent: Buffer for blocking and diluting the Detection Antibody and Enzyme, Substrate: 1X TMB Solution, Certificate of Analysis: Lot-specific instructions for dilution of antibodies and standards, 96 Well Plate: High affinity 96-well plate (included with product Cat. Nos. ending in suffixes -22, -76, -86), Purified antibody, Detection Antibody: Pre-titrated, HRP-conjugated antibody, Standard: Recombinant protein for generating standard curve and calibrating samples, 10X Coating Buffer: Buffer for plating the Capture Antibody, 5X ELISA/ELISPOT Diluent: Buffer for blocking and diluting the Detection Antibody and Enzyme, Substrate: 1X TMB Solution, Certificate of Analysis: Lot-specific instructions for dilution of antibodies and standards, 96 Well Plate: High affinity 96-well plate (included with product Cat. Nos. ending in suffixes -22, -76, -86)
RUO
2-8°C
IL-17A
24 hr 30 min

Spécifications

Q62386
8 pg/mL
HRP
ELISA
Mouse
Absorbance Microplate Reader
16171
Capture Antibody: Pre-titrated, purified antibody, Detection Antibody: Pre-titrated, HRP-conjugated antibody, Standard: Recombinant protein for generating standard curve and calibrating samples, 10X Coating Buffer: Buffer for plating the Capture Antibody, 5X ELISA/ELISPOT Diluent: Buffer for blocking and diluting the Detection Antibody and Enzyme, Substrate: 1X TMB Solution, Certificate of Analysis: Lot-specific instructions for dilution of antibodies and standards, 96 Well Plate: High affinity 96-well plate (included with product Cat. Nos. ending in suffixes -22, -76, -86), Purified antibody, Detection Antibody: Pre-titrated, HRP-conjugated antibody, Standard: Recombinant protein for generating standard curve and calibrating samples, 10X Coating Buffer: Buffer for plating the Capture Antibody, 5X ELISA/ELISPOT Diluent: Buffer for blocking and diluting the Detection Antibody and Enzyme, Substrate: 1X TMB Solution, Certificate of Analysis: Lot-specific instructions for dilution of antibodies and standards, 96 Well Plate: High affinity 96-well plate (included with product Cat. Nos. ending in suffixes -22, -76, -86)
IL-17, IL-17A, Cytotoxic T-lymphocyte-associated antigen 8, CTLA-8
RUO
2-8°C
IL-17A
24 hr 30 min
8-500 pg/mL
Capture Antibody: Pre-titrated, purified antibody
Detection Antibody: Pre-titrated, biotin-conjugated antibody
Standard: Recombinant cytokine for generating standard curve and calibrating samples
ELISA/ELISPOT Coating Buffer Powder: This uncoated ELISA kit with plates may contain ELISA/ELISPOT Coating Buffer Powder (Reconstitute to 1L with dH20 and filter (0.22 μM)) or 10X PBS ELISA Coating Buffer (Dilute 1 part 10X Buffer into 9 parts dH20).
Assay Diluent: 5X concentrated
Detection enzyme: Pre-titrated Avidin-HRP
Substrate Solution: Tetramethylbenzidine (TMB) Substrate Solution
Certificate of Analysis: Lot-specific instructions for dilution of antibodies and standards
96 Well Plate: Corning Costar 9018 (included with product Cat. Nos. ending in suffixes -22, -44, -76, -86)
Uncoated ELISA Kit with Plates
Serum, Supernatant
ELISA
Ctla8; CTLA-8; cytokine; cytotoxic T-lymphocyte-associated antigen 8; cytotoxic T-lymphocyte-associated protein 8; il 17; Il17; IL-17; IL17A; IL-17a; ILN; Interleukin; interleukin 17; interleukin 17 (cytotoxic T-lymphocyte-associated serine esterase 8); interleukin 17 precursor; interleukin 17A; interleukin-17; Interleukin17A; interleukin-17A; R-IL-17-ACtla8; CTLA-8; cytokine; cytotoxic T-lymphocyte-associated antigen 8; cytotoxic T-lymphocyte-associated protein 8; il 17; Il17; IL-17; IL17A; IL-17a; ILN; Interleukin; interleukin 17; interleukin 17 (cytotoxic T-lymphocyte-associated serine esterase 8); interleukin 17 precursor; interleukin 17A; interleukin-17; Interleukin17A; interleukin-17A; R-IL-17-A;
IL17A
Biotin
10 x 96 Tests
Serum 100 μL, Supernatant 100 μL
Interleukin-17 A, IL-17 AA
1 hr 45 min
missing translation for 'recommendations'

missing translation for 'recommendations'

Videos
FDS
Documents

Documents

Promotions

Promotions