missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Mouse Corticosterone Rapid ELISA Kit

Numéro de catalogue. EELR008
Modifier l'affichage
Click to view available options
Quantity:
96 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Numéro de catalogue. Quantity
EELR008 96 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Numéro de catalogue. EELR008 Fournisseur Invitrogen™ Code fournisseur EELR008
Il en reste null

ELISA

The Mouse Corticosterone (CORT) ELISA quantitates CORT in serum, plasma, urine, saliva. Principle of the method The Mouse Cort solid phase competitive ELISA (enzyme linked immunosorbent assay) is designed to measure the amount of the target bound between the pre coated antigen and the HRP linked antibody specific for Mouse Cort. A target specific antigen has been pre coated in the wells of the supplied microplate. Samples, standards, or controls are then added into these wells, where Mouse Cort in the samples (or standards) competes with the immobilized (capture) antigen for sites on the HRP linked detection antibody. Excess conjugate and unbound sample or standard are washed away from the plate. The substrate solution is added to each well, and the enzyme substrate reaction is terminated by the addition of a stop solution, turning the color yellow. The optical density (OD) is measured spectrophotometrically at a wavelength of 450±2 nm. The intensity of this signal is inversely proportional to the concentration of Mouse Cort present in the original specimen, which is determined by comparing the OD of the samples to the standard curve. Achieve fast and reliable results in just 90 minutes with our Rapid ELISA kit, featuring a streamlined protocol that requires only one wash step. Rigorous validation: Each manufactured lot of this ELISA kit is quality tested for criteria such as sensitivity, specificity, precision, and lot-to-lot consistency. See manual for more...

CORTICOSTERONE is an adrenocortical steroid that has modest but significant activities as a mineralocorticoid and a glucocorticoid. [National Center for Biotechnology Information. PubChem Compound Database; CID=5753].

Spécifications

Assay Range 12.5-800 ng/mL
Assay Sensitivity 4.98 ng/mL
Conjugate HRP
Immunoassay Kit Format Coated ELISA Kit
Product Type ELISA
Sample Type Plasma, Saliva, Serum, Urine
Target Species Mouse
For Use With (Equipment) Absorbance Microplate Reader
Gene Alias 17-Deoxycortisol; C21H30O417-Deoxycortisol; C21H30O4;
Interassay CV <10%
Intraassay CV <10%
Kit Contents Pre-coated 96 well plate, Standard, HRP Conjugate, Standard & Sample Diluent, HRP Conjugate Diluent, Wash Buffer, TMB Substrate, Stop Solution, Plate Sealer, Standard, HRP Conjugate, Standard & Sample Diluent, HRP Conjugate Diluent, Wash Buffer, TMB Substrate, Stop Solution, Plate Sealer
Label or Dye HRP
Quantity 96 Tests
Regulatory Status RUO
Sample Volume Plasma 50 μL, Saliva 50 μL, Serum 50 μL, Urine 50 μL
Shipping Condition Wet or Dry Ice
Storage Requirements Refer to product documentation for component specific storage temperature.
Target Corticosterone
Test Time 15 min
Total Assay Time 1 hr 30 min
Afficher plus de résultats Afficher moins de résultats