missing translation for 'onlineSavingsMsg'
Learn More

Mouse anti-β-Tubulin, MP Biomedicals™

Catalog No. MP08L100029
Change view
Click to view available options
Quantité:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantité
MP08L100029 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MP08L100029 Supplier MP Biomedicals Inc Supplier No. 08L100029

missing translation for 'validMainframeMsg'

Mouse Monoclonal Antibody

  • Mouse anti-β-Tubulin mAb offers high specificity and potency and consistency against β-Tubulin expressed by human, mouse, rat, hamster, monkey, goat, and Chlamydomonas reinhardtii species, which has been experimentally validated in Western blotting (WB), Immunohistochemistry (IHC), Immunoprecipitation (IP) and Immunofluorescence (IF) assays.
  • This gene encodes a beta tubulin protein. This protein forms a dimer with alpha tubulin and acts as a structural component of microtubules.
  • Mutations in this gene cause cortical dysplasia, complex, with other brain malformations 6. Alternative splicing results in multiple splice variants. There are multiple pseudogenes for this gene on chromosomes 1, 6, 7, 8, 9, and 13.

Specifications

Antigène β-Tubulin
Applications Western Blot, Immunohistochemistry, Immunoprecipitation, Immunofluorescence
Classification Monoclonal
Conjugué Unconjugated
Formule In PBS buffer with 0.02% sodium azide, 0.05% BSA, 50% glycerol; pH 7.3 to 7.4
Espèces hôtes Mouse
Immunogène Full-length recombinant protein of human β-Tubulin
Méthode de purification Affinity purification
Quantité 100 μL
Statut réglementaire RUO
Primaire ou secondaire Secondary
Espèces cibles Human, Mouse, Rat, Goat, Algae
Poids moléculaire d’anticorps Observed at 50 kDa
Forme Purified
Isotype IgG
Show More Show Less

Unless specified otherwise, for research or further manufacturing use only, not for direct human use.