missing translation for 'onlineSavingsMsg'
Learn More
Learn More
IgM μ chainMouse anti-Rat, Cy5 , Secondary Antibody, MilliporeSigma™
Description
Mouse anti-Rat IgM μ chain Antibody, Cy5 conjugate detects level of IgM μ chain and has been published and validated for use in ELISA and immunofluoresence.
Spécifications
Spécifications
| Antigène | Mouse anti-Rat IgM μ chain |
| Applications | ELISA, Immunofluorescence |
| Classification | Monoclonal |
| Conjugué | Cyanine5 |
| Formule | Purified by affinity chromatography. Liquid in 1.0 mL of PBS/NaN3. |
| Espèces hôtes | Mouse |
| Immunogène | Prepared from rat IgM μ chain. |
| Méthode de purification | Affinity chromatography |
| Quantité | 0.1 mg |
| Primaire ou secondaire | Secondary |
| Afficher plus de résultats |
For Research Use Only