missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mouse anti-Rat IgM Heavy Chain Secondary Antibody (HIS40), R&D Systems™
Description
IgM Heavy Chain Monoclonal specifically detects IgM Heavy Chain in Rat samples. It is validated for Flow Cytometry, Immunohistochemistry, Immunohistochemistry (Paraffin).
Spécifications
Spécifications
| Antigène | IgM Heavy Chain |
| Applications | Flow Cytometry, Immunohistochemistry, Immunohistochemistry (Paraffin) |
| Classification | Monoclonal |
| Clone | HIS40 |
| Concentration | 0.5 mg/mL |
| Conjugué | Unconjugated |
| Dilution | Flow Cytometry, Immunohistochemistry, Immunohistochemistry-Paraffin |
| Alias du gène | Constant region of heavy chain of IgM, Ig Mu Chain C Region BOT, Ig Mu Chain C Region GAL, Ig Mu Chain C Region OU, immunoglobulin heavy constant mu, Immunoglobulin mu chain |
| Symboles de gène | IGHM |
| Espèces hôtes | Mouse |
| Afficher plus de résultats |
For Research Use Only