missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Mouse anti-Rat IgE Secondary Antibody, FITC

Numéro de catalogue. PIMA516812
Modifier l'affichage
Click to view available options
Quantité:
500 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Numéro de catalogue. Quantité
PIMA516812 500 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Numéro de catalogue. PIMA516812 Fournisseur Invitrogen™ Code fournisseur MA516812

Pour de l'aide SVP utiliser la function de clavardage; nous envoyer un couriel à help@thermofisher.com ou appelez le service à la clientèle au 1-800-234-7437.

Mouse Monoclonal Secondary Antibody

For FACS analysis, use 50 μL of the suggested working dilution to label 2x10^7 cells in 100 μL. Mouse anti Rat IgE antibody, clone MARE-1 recognizes rat epsilon heavy chain of immunoglobulin IgE and does not cross-react with other classes of rat immunoglobulin.

Anti-Rat secondary antibodies are affinity-purified antibodies with well-characterized specificity for rat immunoglobulins and are useful in the detection, sorting or purification of its specified target. Secondary antibodies offer increased versatility enabling users to use many detection systems (e.g. HRP, AP, fluorescence). They can also provide greater sensitivity through signal amplification as multiple secondary antibodies can bind to a single primary antibody. Most commonly, secondary antibodies are generated by immunizing the host animal with a pooled population of immunoglobulins from the target species and can be further purified and modified (i.e. immunoaffinity chromatography, antibody fragmentation, label conjugation, etc.) to generate highly specific reagents.

Spécifications

Antigène Rat IgE
Applications Flow Cytometry
Classification Monoclonal
Clone MARE-1
Concentration 1 mg/mL
Conjugué FITC
Formule PBS with 50% glycerol and 0.1% sodium azide
Espèces hôtes Mouse
Immunogène Rat IgE myeloma proteins (IR162, IR1016, IR2 and IR410).
Méthode de purification Affinity chromatography
Quantité 500 μg
Statut réglementaire RUO
Primaire ou secondaire Secondary
Espèces cibles Rat
Contenu et stockage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. Store in the dark.
Type de produit Secondary Antibody
Forme Liquid
Isotype IgG1
Afficher plus de résultats Afficher moins de résultats