missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Mouse anti-Rat IgD Secondary Antibody, Biotin

Numéro de catalogue. PIMA516809
Modifier l'affichage
Click to view available options
Quantité:
500 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Numéro de catalogue. Quantité
PIMA516809 500 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Numéro de catalogue. PIMA516809 Fournisseur Invitrogen™ Code fournisseur MA516809

Pour de l'aide SVP utiliser la function de clavardage; nous envoyer un couriel à help@thermofisher.com ou appelez le service à la clientèle au 1-800-234-7437.

Mouse Monoclonal Secondary Antibody

Mouse anti-Rat IgD Secondary Antibody for ELISA

Anti-Rat secondary antibodies are affinity-purified antibodies with well-characterized specificity for rat immunoglobulins and are useful in the detection, sorting or purification of its specified target. Secondary antibodies offer increased versatility enabling users to use many detection systems (e.g. HRP, AP, fluorescence). They can also provide greater sensitivity through signal amplification as multiple secondary antibodies can bind to a single primary antibody. Most commonly, secondary antibodies are generated by immunizing the host animal with a pooled population of immunoglobulins from the target species and can be further purified and modified (i.e. immunoaffinity chromatography, antibody fragmentation, label conjugation, etc.) to generate highly specific reagents.

Spécifications

Antigène Rat IgD
Applications ELISA
Classification Monoclonal
Clone MARD-3
Concentration 1 mg/mL
Conjugué Biotin
Formule PBS with 50% glycerol and 0.1% sodium azide
Espèces hôtes Mouse
Immunogène IR731 myeloma protein.
Méthode de purification Affinity chromatography
Quantité 500 μg
Statut réglementaire RUO
Primaire ou secondaire Secondary
Espèces cibles Rat
Contenu et stockage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Type de produit Secondary Antibody
Forme Liquid
Isotype IgG1
Afficher plus de résultats Afficher moins de résultats