missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Mouse anti-Rat Ig Mix Secondary Antibody

Numéro de catalogue. PISA510347
Modifier l'affichage
Click to view available options
Quantité:
100 μg
500 μg
1 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Numéro de catalogue. Quantité
PISA510347 100 μg
01671619 500 μg
PISA510346 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Numéro de catalogue. PISA510347 Fournisseur Invitrogen™ Code fournisseur SA510347

Pour de l'aide SVP utiliser la function de clavardage; nous envoyer un couriel à help@thermofisher.com ou appelez le service à la clientèle au 1-800-234-7437.

Mouse Monoclonal Secondary Antibody

SA1-10652 detects rat immunoglobulin. SA1-10652 has been successfully used in ELISA applications. The SA1-10652 immunogen is purified rat immunoglobulin.

Anti-Rat secondary antibodies are affinity-purified antibodies with well-characterized specificity for rat immunoglobulins and are useful in the detection, sorting or purification of its specified target. Secondary antibodies offer increased versatility enabling users to use many detection systems (e.g. HRP, AP, fluorescence). They can also provide greater sensitivity through signal amplification as multiple secondary antibodies can bind to a single primary antibody. Most commonly, secondary antibodies are generated by immunizing the host animal with a pooled population of immunoglobulins from the target species and can be further purified and modified (i.e. immunoaffinity chromatography, antibody fragmentation, label conjugation, etc.) to generate highly specific reagents.

Spécifications

Antigène Rat Ig Mix
Applications ELISA
Classification Monoclonal
Clone RATIG12-306.21
Concentration 0.6 mg/mL
Conjugué Unconjugated
Formule PBS with no preservative; pH 7.4
Espèces hôtes Mouse
Immunogène Purified rat immunoglobulin.
Méthode de purification Protein G
Quantité 100 μg
Statut réglementaire RUO
Primaire ou secondaire Secondary
Espèces cibles Rat
Contenu et stockage -20°C, Avoid Freeze/Thaw Cycles
Type de produit Secondary Antibody
Forme Liquid
Isotype IgG1
Afficher plus de résultats Afficher moins de résultats