missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Light chain Mouse anti-Rabbit, Cy3 , Secondary Antibody, MilliporeSigma™
Description
Mouse anti-Rabbit light chain Antibody, Cy3 conjugate detects level of Rabbit light chain and has been published and validated for use in ELISA and western blotting.
Spécifications
Spécifications
| Antigène | Mouse anti-Rabbit light chain |
| Applications | ELISA, Western Blot |
| Classification | Monoclonal |
| Conjugué | Cyanine3 |
| Formule | Isolated from ascites by chromatographic procedures. Liquid in 0.01M Sodium Phosphate, 0.25M NaCl, pH 7.6, 0.05% Sodium Azide. 15mg/mL BSA as stabilizer. |
| Espèces hôtes | Mouse |
| Immunogène | Prepared from rabbit IgG light chain. |
| Méthode de purification | Ascites |
| Quantité | 0.5 mg |
| Primaire ou secondaire | Secondary |
| Afficher plus de résultats |
For Research Use Only