missing translation for 'onlineSavingsMsg'
Learn More

Light chain Mouse anti-Rabbit, Alkaline Phosphatase , Secondary Antibody, MilliporeSigma™

Numéro de catalogue. MAB201AMI
Modifier l'affichage
Click to view available options
Quantité:
0.5 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Numéro de catalogue. Quantité
MAB201AMI 0.5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Numéro de catalogue. MAB201AMI Fournisseur MilliporeSigma Code fournisseur MAB201A

Pour de l'aide SVP utiliser la function de clavardage; nous envoyer un couriel à help@thermofisher.com ou appelez le service à la clientèle au 1-800-234-7437.

Mouse Monoclonal Antibody

This Mouse anti-Rabbit light chain Antibody, Alkaline Phosphatase conjugate is validated for use in ELISA, western blotting for the detection of Rabbit light chain.

Spécifications

Antigène Mouse anti-Rabbit light chain
Applications ELISA, Western Blot
Classification Monoclonal
Conjugué Alkaline Phosphatase (AP)
Formule Isolated from ascites by chromatographic procedures. Liquid in 0.01M Tris-HCl, 0.25M NaCl, pH 8.0 with 0.05% Sodium Azide. 15mg/mL BSA as stabilizer.
Espèces hôtes Mouse
Immunogène Prepared from rabbit IgG light chain.
Méthode de purification Ascites
Quantité 0.5 mL
Primaire ou secondaire Secondary
Espèces cibles Rabbit
Contenu et stockage 2-8°C lyophilized for up to 12 months −20°C in undiluted aliquots for up to 12 months. Keep away from light.
Forme Purified
Isotype IgG
Afficher plus de résultats Afficher moins de résultats

For Research Use Only