missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mouse anti-Rabbit IgM (H+L) Secondary Antibody (NRBM) [Alexa Fluor™ Plus 488], R&D Systems™
Description
IgM (H+L) Monoclonal antibody specifically detects IgM (H+L) in Rabbit samples. It is validated for Flow Cytometry, Immunohistochemistry, Immunohistochemistry (Frozen)
Spécifications
Spécifications
| Antigène | IgM (H+L) |
| Applications | Flow Cytometry, Immunohistochemistry, Immunohistochemistry (Frozen) |
| Classification | Monoclonal |
| Clone | NRBM |
| Conjugué | Alexa Fluor Plus 488 |
| Formule | 50mM Sodium Borate |
| Espèces hôtes | Mouse |
| Immunogène | Rabbit IgM |
| Méthode de purification | Protein A purified |
| Quantité | 0.1 mL |
| Afficher plus de résultats |