missing translation for 'onlineSavingsMsg'
Learn More

IgG Mouse anti-Rabbit, HRP, Polyclonal, Secondary Antibody, MilliporeSigma™

Numéro de catalogue. AP188PMI
Modifier l'affichage
Click to view available options
Quantité:
1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Numéro de catalogue. Quantité
AP188PMI 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Numéro de catalogue. AP188PMI Fournisseur MilliporeSigma Code fournisseur AP188P

Pour de l'aide SVP utiliser la function de clavardage; nous envoyer un couriel à help@thermofisher.com ou appelez le service à la clientèle au 1-800-234-7437.

Mouse Polyclonal Antibody

Mouse anti-Rabbit IgG Antibody, HRP conjugate, Species Adsorbed detects level of Rabbit IgG and has been published and validated for use in ELISA and western blotting.

Spécifications

Antigène Mouse anti-Rabbit IgG Species adsorbed
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 0.8mg/mL
Conjugué HRP
Formule Lyophilized. Buffer = 0.01M Sodium Phosphate, 0.25M NaCl, pH 7.6 with 15mg/mL BSA and 0.01% thimerosal. RECONSTITUTION: Reconstitute with 1000μL of sterile distilled water. Centrifuge product if it is not completely clear after standing 1-2 hour
Espèces hôtes Mouse
Quantité 1 mL
Primaire ou secondaire Secondary
Espèces cibles Rabbit
Contenu et stockage 2-8°C lyophilized for up to 12 months −20°C in undiluted aliquots for up to 12 months. Avoid repeated freeze/thaw cycles.
Forme Lyophilized
Afficher plus de résultats Afficher moins de résultats

For Research Use Only