missing translation for 'onlineSavingsMsg'
Learn More

Mouse Anti-Rabbit IgG Antibody [6C1A8], ProSci

Numéro de catalogue. 16595720
Modifier l'affichage
Click to view available options
Quantité:
1.0 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Numéro de catalogue. Quantité
16595720 1.0 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Numéro de catalogue. 16595720 Fournisseur ProSci Llc Code fournisseur PM62851000

Pour de l'aide SVP utiliser la function de clavardage; nous envoyer un couriel à help@thermofisher.com ou appelez le service à la clientèle au 1-800-234-7437.

Mouse Control Antibody

Mouse Anti-Rabbit IgG Antibody specifically detects Mouse Anti-Rabbit IgG in Rabbit samples. It is validated for ELISA, Western Blot, Immunohistochemistry

Spécifications

Antigène Mouse Anti-Rabbit IgG
Applications ELISA, Western Blot, Immunohistochemistry
Classification Monoclonal
Clone 6C1A8
Concentration 1 mg/mL
Conjugué Unconjugated
Dilution Immunoblotting: 1/5,000 - 1/50,000, ELISA: 1/5,000 - 1/50,000, Immunohistochemistry: 1/100 - 1/1,000
Formule PBS pH 7.2, 1% BSA, 0.02% Thimerosal
Espèces hôtes Mouse
Quantité 1.0 mg
Statut réglementaire RUO
Primaire ou secondaire Secondary
Spécifications de test No cross reactivity with immunoglobulins from other species.
Espèces cibles Rabbit
Contenu et stockage Antibody can be stored at -20°C, stable for up to one year. As with all antibodies care should be taken to avoid repeated freeze thaw cycles. Antibodies should not be exposed to prolonged high temperatures.
Type de produit Antibody
Forme Liquid
Isotype IgG2
Afficher plus de résultats Afficher moins de résultats