missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mouse anti-Human IgG1 Fc Secondary Antibody (2C11cc) [Janelia Fluor 525], R&D Systems™
Description
IgG1 Fc Monoclonal antibody specifically detects IgG1 Fc in Human samples. It is validated for ELISA, Immunohistochemistry, Immunohistochemistry (Frozen), Immunodiffusion
Spécifications
Spécifications
| Antigène | IgG1 Fc |
| Applications | ELISA, Immunohistochemistry, Immunohistochemistry (Frozen), Immunodiffusion |
| Classification | Monoclonal |
| Clone | 2C11cc |
| Conjugué | Janelia Fluor 525 |
| Formule | 50mM Sodium Borate |
| Espèces hôtes | Mouse |
| Immunogène | Hybridoma clone has been derived from hybridization of Sp2/0 myeloma cells with spleen cells of Balb/c mice immunized with IgG from human serum. |
| Méthode de purification | Protein A purified |
| Quantité | 0.2 mL |
| Afficher plus de résultats |