missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mouse anti-Human IgG (H+L) Secondary Antibody (4D2D9G8), Allophycocyanin, R&D Systems™
Description
IgG (H+L) Monoclonal specifically detects IgG (H+L) in Human samples. It is validated for ELISA, Western Blot.
Spécifications
Spécifications
| Antigène | IgG (H+L) |
| Applications | ELISA, Western Blot |
| Classification | Monoclonal |
| Clone | 4D2D9G8 |
| Conjugué | APC |
| Dilution | Western Blot : 1:500 - 1:2000, ELISA : 1:10000 |
| Formule | PBS |
| Espèces hôtes | Mouse |
| Immunogène | Human IgG was isolated from human sera and purified by chromatography. |
| Méthode de purification | Protein G purified |
| Afficher plus de résultats |