missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Mouse anti-Human IgG (H+L) Secondary Antibody
Description
MA5-14728 targets Human IgG in ELISA, IP, and RIA applications and shows reactivity with Human samples. The MA5-14728 immunogen is human Serum IgG. Product MA514728 is a smaller package size of MII0303 (formerly sold as a Seradyn product).
Anti-Human secondary antibodies are affinity-purified antibodies with well-characterized specificity for human immunoglobulins and are useful in the detection, sorting or purification of its specified target. Secondary antibodies offer increased versatility enabling users to use many detection systems (e.g. HRP, AP, fluorescence). They can also provide greater sensitivity through signal amplification as multiple secondary antibodies can bind to a single primary antibody. Most commonly, secondary antibodies are generated by immunizing the host animal with a pooled population of immunoglobulins from the target species and can be further purified and modified (i.e. immunoaffinity chromatography, antibody fragmentation, label conjugation, etc.) to generate highly specific reagents.
Spécifications
Spécifications
| Antigène | Human IgG (H+L) |
| Applications | ELISA, Immunoprecipitation, Radioimmune Assays (RIA) |
| Classification | Monoclonal |
| Clone | F37-5 |
| Concentration | 1 mg/mL |
| Conjugué | Unconjugated |
| Formule | PBS with 1mg/mL BSA and 0.05% sodium azide; pH 7.4 |
| Espèces hôtes | Mouse |
| Immunogène | Human Serum IgG. |
| Méthode de purification | Ion-exchange chromatography |
| Afficher plus de résultats |