missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mouse anti-Human IgG Fc Secondary Antibody (MK 1 A6) [Janelia Fluor 669], R&D Systems™
Description
IgG Fc Monoclonal antibody specifically detects IgG Fc in Human samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry (Frozen)
Spécifications
Spécifications
| Antigène | IgG Fc |
| Applications | Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry (Frozen) |
| Classification | Monoclonal |
| Clone | MK 1 A6 |
| Conjugué | Janelia Fluor 669 |
| Formule | 50mM Sodium Borate |
| Espèces hôtes | Mouse |
| Immunogène | Human IgG Polyclonal |
| Méthode de purification | Protein A purified |
| Quantité | 0.2 mL |
| Afficher plus de résultats |