missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Mouse anti-Human IgG Fc Secondary Antibody, Biotin

Numéro de catalogue. 054240
Encompass
Modifier l'affichage
Click to view available options
Quantité:
500 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Numéro de catalogue. Quantité
054240 500 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Numéro de catalogue. 054240 Fournisseur Invitrogen™ Code fournisseur 054240

Pour de l'aide SVP utiliser la function de clavardage; nous envoyer un couriel à help@thermofisher.com ou appelez le service à la clientèle au 1-800-234-7437.

Mouse Monoclonal Secondary Antibody

Reconstitute with 0.5 mL of distilled water.

Anti-Human secondary antibodies are affinity-purified antibodies with well-characterized specificity for human immunoglobulins and are useful in the detection, sorting or purification of its specified target. Secondary antibodies offer increased versatility enabling users to use many detection systems (e.g. HRP, AP, fluorescence). They can also provide greater sensitivity through signal amplification as multiple secondary antibodies can bind to a single primary antibody. Most commonly, secondary antibodies are generated by immunizing the host animal with a pooled population of immunoglobulins from the target species and can be further purified and modified (i.e. immunoaffinity chromatography, antibody fragmentation, label conjugation, etc.) to generate highly specific reagents.

Spécifications

Antigène Human IgG Fc
Applications ELISA
Classification Monoclonal
Clone HP6017
Conjugué Biotin
Formule PBS with 1% BSA and 0.1% sodium azide; pH 7.4
Espèces hôtes Mouse
Immunogène Gamma Immunoglobin.
Méthode de purification Purified
Quantité 500 μL
Statut réglementaire RUO
Primaire ou secondaire Secondary
Espèces cibles Human
Contenu et stockage 4°C
Type de produit Secondary Antibody
Forme Lyophilized
Isotype IgG2a κ
Afficher plus de résultats Afficher moins de résultats