missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mouse anti-Human IgG F(ab) Secondary Antibody (4A11) [Janelia Fluor 669], R&D Systems™
Description
IgG F(ab) Monoclonal antibody specifically detects IgG F(ab) in Human samples. It is validated for Western Blot, Flow Cytometry, ELISA
Spécifications
Spécifications
| Antigène | IgG F(ab) |
| Applications | Western Blot, Flow Cytometry, ELISA |
| Classification | Monoclonal |
| Clone | 4A11 |
| Conjugué | Janelia Fluor 669 |
| Formule | 50mM Sodium Borate |
| Espèces hôtes | Mouse |
| Immunogène | Purified human IgG. |
| Méthode de purification | Protein A purified |
| Quantité | 0.1 mL |
| Afficher plus de résultats |