missing translation for 'onlineSavingsMsg'
Learn More
Learn More
IgG Mouse anti-Goat, HRP, Polyclonal, Secondary Antibody, MilliporeSigma™
Description
This Mouse anti-Goat IgG Antibody, HRP conjugate, Species Adsorbed is validated for use in ELISA, western blotting for the detection of Goat IgG.
Spécifications
Spécifications
| Antigène | Mouse anti-Goat IgG Species adsorbed |
| Applications | ELISA, Western Blot |
| Classification | Polyclonal |
| Conjugué | HRP |
| Espèces hôtes | Mouse |
| Quantité | 1 mL |
| Primaire ou secondaire | Secondary |
| Espèces cibles | Goat |
For Research Use Only