Learn More
Invitrogen™ Mouse anti-Goat IgG (H+L) Cross-Adsorbed Secondary Antibody
Description
Concentration may vary slightly from lot-to-lot, see lot-specific datasheet for exact concentration. Product # 31107 has been successfully used in Western blot, IF, ICC, IHC, IP and FACS applications. Product # 31107 reacts with the heavy chains of goat IgG and with the light chains common to most goat immunoglobulins, but does not react against non-immunoglobulin serum proteins. The antibody has been tested by ELISA and/or solid-phase adsorbed to ensure minimal cross-reaction with mouse, human and rabbit serum proteins. However, this antibody may cross-react with immunoglobulins from other species. Store product at 4°C until opened. After opening, dilute only enough antibody for a single day's use. Store remainder at 4°C under sterile conditions. Country of origin: USA.
Spécifications
Spécifications
| Antigène | Goat IgG (H+L) Cross-Adsorbed |
| Applications | Flow Cytometry, Immunohistochemistry, Immunoprecipitation, Western Blot, Immunocytochemistry |
| Classification | Polyclonal |
| Concentration | 1.8 mg/mL |
| Conjugué | Unconjugated |
| Formule | PBS with no preservative; pH 7.6 |
| Espèces hôtes | Mouse |
| Méthode de purification | Antigen affinity chromatography |
| Quantité | 1.5 mg |
| Statut réglementaire | RUO |
| Afficher plus de résultats |