missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Mouse anti-Goat IgG (H+L) Cross-Adsorbed Secondary Antibody, HRP

Numéro de catalogue. PI31400
Modifier l'affichage
Click to view available options
Quantité:
1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Numéro de catalogue. Quantité
PI31400 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Numéro de catalogue. PI31400 Fournisseur Invitrogen™ Code fournisseur 31400
  • ONLINE EXCLUSIVE PRICE
    $562.48
    $327.00 / chaque

    Économisez $235.48

    42.0 % de rabais Les écarts et divulgations suivants peuvent entraîner des prix inférieurs aux rabais indiqués, sans toutefois entraîner de réductions de prix supplémentaires : Le prix affiché reflète une réduction promotionnelle temporaire. Aucun achat additionnel ni engagement commercial n’est requis pour bénéficier du prix réduit. Le prix peut être modifié en tout temps. Le prix promotionnel est valide uniquement pour les commandes en ligne. Votre tarification contractuelle peut différer. D’autres exclusions peuvent s’appliquer.
Afficher toutes les promotions

Pour de l'aide SVP utiliser la function de clavardage; nous envoyer un couriel à help@thermofisher.com ou appelez le service à la clientèle au 1-800-234-7437.

Mouse Polyclonal Secondary Antibody

Concentration may vary slightly from lot-to-lot, see lot-specific datasheet for exact concentration. This antibody has been successfully used in Western blot, and ICC applications. Antibody Specificity: This antibody reacts with the heavy chains of goat IgG and with light chains common to most goat immunoglobulins. The antibody exhibits minimal inherent cross-reactivity to mouse serum proteins. No antibody was detected against non-immunoglobulin serum proteins. The antibody has been tested by ELISA and/or solid-phase adsorbed to ensure minimal cross-reaction with mouse, human and rabbit serum proteins. However, this antibody may cross-react with immunoglobulins from other species. Restoration and Storage: Store product at 2-8 °C until opened. Restore with 1.0 mL distilled water (0.8 mg/mL after restoration). Centrifuge product if it is not completely clear after standing for 1-2 hours at room temperature. To judge clarity, draw product into a pasteur pipette. Product may be stored for several weeks at 4°C as an undiluted liquid. After dilution, do not use for more than one day. To extend the shelf-life of this product, add an equal volume of glycerol to make a final concentration of approximately 50% glycerol and store at -20°C. Country of Origin: USA.

Anti-Goat secondary antibodies are affinity-purified antibodies with well-characterized specificity for goat immunoglobulins and are useful in the detection, sorting or purification of its specified target. Secondary antibodies offer increased versatility enabling users to use many detection systems (e.g. HRP, AP, fluorescence). They can also provide greater sensitivity through signal amplification as multiple secondary antibodies can bind to a single primary antibody. Most commonly, secondary antibodies are generated by immunizing the host animal with a pooled population of immunoglobulins from the target species and can be further purified and modified (i.e. immunoaffinity chromatography, antibody fragmentation, label conjugation, etc.) to generate highly specific reagents.

Spécifications

Antigène Goat IgG (H+L) Cross-Adsorbed
Applications Immunohistochemistry, Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.8 mg/mL
Conjugué HRP
Formule PBS with 15mg/mL BSA and no preservative; pH 7.6
Espèces hôtes Mouse
Méthode de purification Antigen affinity chromatography
Quantité 1 mL
Statut réglementaire RUO
Primaire ou secondaire Secondary
Espèces cibles Goat
Contenu et stockage 4°C
Type de produit Secondary Antibody
Forme Lyophilized
Isotype IgG
Afficher plus de résultats Afficher moins de résultats