missing translation for 'onlineSavingsMsg'
Learn More

Mouse Anti-Goat IgG Antibody [1G12H5] (HRP), ProSci

Numéro de catalogue. 16916761
Modifier l'affichage
Click to view available options
Quantité:
0.1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Numéro de catalogue. Quantité
16916761 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Numéro de catalogue. 16916761 Fournisseur ProSci Llc Code fournisseur 16030100

Pour de l'aide SVP utiliser la function de clavardage; nous envoyer un couriel à help@thermofisher.com ou appelez le service à la clientèle au 1-800-234-7437.

Mouse Control Antibody

Mouse Anti-Goat IgG Antibody specifically detects Mouse Anti-Goat IgG in Goat samples. It is validated for ELISA, Western Blot, Immunohistochemistry

Spécifications

Antigène Mouse Anti-Goat IgG
Applications ELISA, Western Blot, Immunohistochemistry
Classification Monoclonal
Clone 1G12H5
Concentration 1 mg/mL
Conjugué HRP
Dilution Immunoblotting: 1/5,000 - 1/50,000, ELISA: 1/5,000 - 1/50,000, Immunohistochemistry: 1/100 - 1/1,000
Formule PBS pH 7.2, 1% BSA, 0.02% Thimerosal
Espèces hôtes Mouse
Quantité 0.1 mg
Statut réglementaire RUO
Primaire ou secondaire Secondary
Spécifications de test No cross reactivity with immunoglobulins from other species.
Espèces cibles Goat
Contenu et stockage Store at 2-8°C. The product is stable for about 4-6 weeks at 4°C as an undiluted liquid. For extended storage, aliquot and freeze the product at -70°C or below. Avoid repeated freezing and thawing. Or alternatively, add an equal volume of glycerol for a final concentration of 50% and store at -20°C.
Type de produit Antibody
Forme Liquid
Isotype IgG2
Afficher plus de résultats Afficher moins de résultats