missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Mouse anti-Feline IgE Secondary Antibody

Numéro de catalogue. PISA510296
Modifier l'affichage
Click to view available options
Quantité:
1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Numéro de catalogue. Quantité
PISA510296 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Numéro de catalogue. PISA510296 Fournisseur Invitrogen™ Code fournisseur SA510296

Pour de l'aide SVP utiliser la function de clavardage; nous envoyer un couriel à help@thermofisher.com ou appelez le service à la clientèle au 1-800-234-7437.

Mouse Monoclonal Secondary Antibody

Mouse anti-Feline IgE Secondary Antibody for ELISA, Western Blot

Anti-Feline secondary antibodies are affinity-purified antibodies with well-characterized specificity for feline immunoglobulins and are useful in the detection, sorting or purification of its specified target. Secondary antibodies offer increased versatility enabling users to use many detection systems (e.g. HRP, AP, fluorescence). They can also provide greater sensitivity through signal amplification as multiple secondary antibodies can bind to a single primary antibody. Most commonly, secondary antibodies are generated by immunizing the host animal with a pooled population of immunoglobulins from the target species and can be further purified and modified (i.e. immunoaffinity chromatography, antibody fragmentation, label conjugation, etc.) to generate highly specific reagents.

Spécifications

Antigène Feline IgE
Applications ELISA, Western Blot
Classification Monoclonal
Clone 2C1
Concentration 3.25 mg/mL
Conjugué Unconjugated
Formule PBS with 0.09% sodium azide; pH 7.2
Espèces hôtes Mouse
Immunogène Feline IgE.
Méthode de purification Protein A
Quantité 1 mg
Statut réglementaire RUO
Primaire ou secondaire Secondary
Espèces cibles Feline
Contenu et stockage -20°C
Type de produit Secondary Antibody
Forme Liquid
Isotype IgG2b κ
Afficher plus de résultats Afficher moins de résultats