Learn More
Invitrogen™ Mouse anti-Feline Ig Mix Secondary Antibody
Mouse Monoclonal Secondary Antibody
Supplier:  Invitrogen™ SA510359
 Afficher plus de versions de ce produit
Numéro de catalogue. PISA510359
Il en reste null
Description
Description
SA1-10641 detects feline Immunoglobulin. Does not react with bovine, equine, canine, rabbit, sheep, porcine, donkey, guinea pig, goat or human. SA1-10641 has been successfully used in ELISA applications. The SA1-10641 immunogen is purified feline immunoglobulin.
Anti-Feline secondary antibodies are affinity-purified antibodies with well-characterized specificity for feline immunoglobulins and are useful in the detection, sorting or purification of its specified target. Secondary antibodies offer increased versatility enabling users to use many detection systems (e.g. HRP, AP, fluorescence). They can also provide greater sensitivity through signal amplification as multiple secondary antibodies can bind to a single primary antibody. Most commonly, secondary antibodies are generated by immunizing the host animal with a pooled population of immunoglobulins from the target species and can be further purified and modified (i.e. immunoaffinity chromatography, antibody fragmentation, label conjugation, etc.) to generate highly specific reagents.
Spécifications
Identifiants chimiques
Feline Ig Mix
1.0 mg/mL
ELISA
Unconjugated
Mouse
RUO
PBS with no preservative; pH 7.4
Secondary
Protein A
FIG10-102.8
-20°C, Avoid Freeze/Thaw Cycles
Monoclonal
Liquid
IgG1
Feline
Purified feline immunoglobulin.
Secondary Antibody
Spécifications
Feline Ig Mix
Monoclonal
1.0 mg/mL
PBS with no preservative; pH 7.4
Purified feline immunoglobulin.
1 mg
Secondary
-20°C, Avoid Freeze/Thaw Cycles
Liquid
ELISA
FIG10-102.8
Unconjugated
Mouse
Protein A
RUO
Feline
Secondary Antibody
IgG1
missing translation for 'recommendations'
missing translation for 'recommendations'
Videos
FDS