missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mouse anti-Canine IgA/IgG/IgM (H+L) Secondary Antibody (K99 2E3) [Janelia Fluor 669], R&D Systems™
Description
IgA/IgG/IgM (H+L) Monoclonal antibody specifically detects IgA/IgG/IgM (H+L) in Canine samples. It is validated for ELISA
Spécifications
Spécifications
| Antigène | IgA/IgG/IgM (H+L) |
| Applications | ELISA |
| Classification | Monoclonal |
| Clone | K99 2E3 |
| Conjugué | Janelia Fluor 669 |
| Formule | 50mM Sodium Borate |
| Espèces hôtes | Mouse |
| Immunogène | Canine immunoglobulin |
| Méthode de purification | Protein A purified |
| Quantité | 0.1 mL |
| Afficher plus de résultats |