missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mouse anti-Bovine IgG2 Heavy Chain Secondary Antibody (IL-A73) [Janelia Fluor 525], R&D Systems™
Description
IgG2 Heavy Chain Monoclonal antibody specifically detects IgG2 Heavy Chain in Bovine samples. It is validated for Flow Cytometry, ELISA, Immunoprecipitation
Spécifications
Spécifications
| Antigène | IgG2 Heavy Chain |
| Applications | Flow Cytometry, ELISA, Immunoprecipitation |
| Classification | Monoclonal |
| Clone | IL-A73 |
| Conjugué | Janelia Fluor 525 |
| Formule | 50mM Sodium Borate |
| Espèces hôtes | Mouse |
| Immunogène | Purified bovine IgG2 |
| Méthode de purification | Protein A purified |
| Quantité | 0.1 mL |
| Afficher plus de résultats |