missing translation for 'onlineSavingsMsg'
Learn More
Learn More
United Scientific™ Motion Balls, Solid or Drilled
Motion Balls are ideal for pendulum bobs or collision experiments.
Supplier: United Scientific™ PNBS25
Description
- These drilled balls are excellent for use as pendulum bobs or in impact and collision experiments.
- All drilled balls feature 3 mm countersunk holes so pendulum cord knots can be recessed.
- Constructed from various materials.
Identifiants chimiques
| Steel Ball |
Spécifications
| Steel Ball | |
| Steel | |
| Drilled Balls | |
| 25 mm |