Learn More
United Scientific™ Motion Balls, Solid or Drilled
Motion Balls are ideal for pendulum bobs or collision experiments.
$2.93 - $9.21
Specifications
Product Type Steel Ball
View More Specs
Products 7
Catalog Number Mfr. No. Material Includes Diameter (Metric) Ball Price Quantity & Availability  
Catalog Number Mfr. No. Material Includes Diameter (Metric) Ball Price Quantity & Availability  
S01313
View Documents Promotion Details
United Scientific™
PNBS13S
Steel Solid Balls 13 mm
Each for $4.32
Only null left
S01312
View Documents Promotion Details
United Scientific™
PNBS19S
Steel Solid Balls 19 mm
Each for $3.50
Only null left
S01310
View Documents Promotion Details
United Scientific™
PNBS25S
Steel Solid Balls 25 mm
Each for $8.39
Only null left
S01309
View Documents Promotion Details
United Scientific™
PNBW25
Wood Drilled Balls 25 mm
Each for $2.93
Only null left
S01311
View Documents Promotion Details
United Scientific™
PNBS25
Steel Drilled Balls 25 mm
Each for $9.21
Only null left
S01314
View Documents Promotion Details
United Scientific™
PNBCK25
Cork Drilled Balls 25 mm
Each for $4.82
Only null left
S01315
View Documents Promotion Details
United Scientific™
PNBA25
Aluminum Drilled Balls 25 mm
Each for $7.86
Only null left
Description
Description
  • These drilled balls are excellent for use as pendulum bobs or in impact and collision experiments.
  • All drilled balls feature 3 mm countersunk holes so pendulum cord knots can be recessed.
  • Constructed from various materials.
Specifications
Specifications
Steel Ball