missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Monkey TIMP-1 ELISA Kit
ELISA
$944.00 - $4,215.00
Spécifications
| Accession Number | Clgi; Collagenase inhibitor; collagenase inhibitor 16C8 fibroblast; EG-1; embryogenin-1; EPA; EPO; erythroid potentiating activity; erythroid-potentiating activity; Fibroblast collagenase inhibitor; FLJ90373; HCI; LOC100009047; Metalloproteinase inhibitor; metalloproteinase inhibitor 1; metalloproteinase tissue inhibitor; metalloproteinase tissue inhibitor 1; MMP inhibitor; RP1-230G1.3; TIMP; TIMP 1; TIMP metallopeptidase inhibitor 1; Timp1; TIMP-1; TIMP-1 protein; tissue inhibitor of matrix metalloproteinase-1; tissue inhibitor of metallopeptidase 1; tissue inhibitor of metalloproteinase; tissue inhibitor of metalloproteinase 1; tissue inhibitor of metalloproteinase 1 (erythroid potentiating activity, collagenase inhibitor); tissue inhibitor of metalloproteinase-1; tissue inhibitor of metalloproteinases; tissue inhibitor of metalloproteinases 1; TPA-induced protein; TPA-S1 |
|---|---|
| Assay Range | 12.5 to 3,000 pg/mL |
| Assay Sensitivity | 12.5 pg/mL |
| Conjugate | HRP |
| Product Type | ELISA |
| Numéro de catalogue | Numéro du manufacturier. | Quantity | Prix | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Numéro de catalogue | Numéro du manufacturier. | Quantity | Prix | Quantity & Availability | |||||
EP18RBX5
|
Invitrogen™
EP18RBX5 |
5 x 96 Tests |
chaque for $4,215.00
|
|
Connectez-vous ou enregistrez-vous pour vérifier votre prix et la disponibilité.
|
||||
EP18RB
|
Invitrogen™
EP18RB |
96 Tests |
chaque for $944.00
|
|
Connectez-vous ou enregistrez-vous pour vérifier votre prix et la disponibilité.
|
||||
Description
Monkey TIMP-1 quantitates monkey TIMP-1 in serum, plasma, supernatant. The assay will exclusively recognize both natural and recombinant monkey TIMP-1.
TIMP1 (metalloproteinase inhibitor 1) is a metalloproteinase inhibitor that functions by forms one-to-one complexes with target metallproteinases, such as collagenases, and irreversible inactivates them by binding to their catalytic zinc cofactor. TIMP1 acts on MMP1, MMP2, MMP3, MMP7, MMP8, MMP9, MMP10, MMP11, MMP12, MMP14, and MMP16. It also functions as a growth factor that regulates cell differenation, migration, and cell death. TIMP1 activates cellular signaling cascades via CD63 and ITGB1.Spécifications
| Clgi; Collagenase inhibitor; collagenase inhibitor 16C8 fibroblast; EG-1; embryogenin-1; EPA; EPO; erythroid potentiating activity; erythroid-potentiating activity; Fibroblast collagenase inhibitor; FLJ90373; HCI; LOC100009047; Metalloproteinase inhibitor; metalloproteinase inhibitor 1; metalloproteinase tissue inhibitor; metalloproteinase tissue inhibitor 1; MMP inhibitor; RP1-230G1.3; TIMP; TIMP 1; TIMP metallopeptidase inhibitor 1; Timp1; TIMP-1; TIMP-1 protein; tissue inhibitor of matrix metalloproteinase-1; tissue inhibitor of metallopeptidase 1; tissue inhibitor of metalloproteinase; tissue inhibitor of metalloproteinase 1; tissue inhibitor of metalloproteinase 1 (erythroid potentiating activity, collagenase inhibitor); tissue inhibitor of metalloproteinase-1; tissue inhibitor of metalloproteinases; tissue inhibitor of metalloproteinases 1; TPA-induced protein; TPA-S1 | |
| 12.5 pg/mL | |
| ELISA | |
| Colorimetric Microplate Reader | |
| <10 | |
| HRP | |
| Plasma 50 μL, Serum 50 μL, Supernatant 50 μL | |
| TIMP1 | |
| 4 hrs 45 mins |
| 12.5 to 3,000 pg/mL | |
| HRP | |
| Plasma, Serum, Supernatant | |
| <12% | |
| Pre-coated 96 well plate, Standard, Assay Diluent concentrate, Biotinylated Detection Antibody, SAV-HRP, Wash Buffer, Chromogen, Stop Solution, Adhesive Plate Covers | |
| RUO | |
| 2°C to 8°C | |
| 1 hrs 20 mins |