missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Molybdenum AA Standard, 1000 ppm Mo in 3% HCl/tr HF, Ricca Chemical
Specifications
Specifications
| Packaging | Natural Poly Bottle |
| Concentration or Composition Notes | 1,000ppm |
| CAS Min % | 94.0, 3.9, 0.14, 0.01 |
| CAS Max % | 98.0, 4.06, 0.15, 0.01 |
| Physical Form | Liquid |
| Color | Colorless |
| Boiling Point | 100°C |
| Type | Single-element Standard Solution |
| Solvent or Matrix | 3% HCl/tr HF |
| Quantity | 500 mL |
| Show More |