accessibility menu, dialog, popup

UserName

OMoOO

CAS RN 1313-27-5

IUPAC Name: trioxomolybdenum
Synonyms: molybdenum trioxide|MoO3
Molecular Weight (g/mol): 143.95
Molecular Formula: MoO3
InChi Key: JKQOBWVOAYFWKG-UHFFFAOYSA-N
SMILES: O=[Mo](=O)=O
Quantity 
  • (7)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (5)
Physical Form 
  • (8)
  • (2)
  • (3)
Percent Purity 
  • (1)
  • (4)
  • (2)
  • (3)
  • (2)
Grade 
  • (2)
  • (2)
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (16)
  • (7)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (5)
  • (8)
  • (2)
  • (3)
  • (1)
  • (4)
  • (2)
  • (3)
  • (2)
  • (2)
  • (2)
  • (1)
  • (2)
  • (1)