missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Epidermal Growth Factor, Biotinylated, complexed to Alexa Fluor™ 647 Streptavidin (Alexa Fluor™ 647 EGF complex)

Catalog No. E35351
Encompass
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
E35351 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. E35351 Supplier Invitrogen™ Supplier No. E35351

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Complexes of the dye-streptavidin molecule and biotinylated EGF

Fluorescently labeled EGF has enabled scientists to investigate receptor-membrane interactions, study receptor distribution, calculate rate constants for the interaction of EGF with its receptor, and more.

  • Biotinylated conjugates utilize biotin-XX, which contains long spacer arm designed to enhance probe affinity for EGF receptor
  • EGF molecule: 53 amino acids, MW=6,045 Da
  • Number of fluorophore molecules on each EGF molecule: 1
  • Fluorescence is typically monitored using a flow cytometer, fluorescence microscope, or fluorimeter

Cell Analysis, Cell Viability, Proliferation and Function, Cellular Imaging, Endocytosis, Exocytosis and Phagocytosis, Flow Cytometry, Flow Cytometry Endocytosis, Exocytosis and Phagocytosis Assays, Flow Cytometry of Cellular Processes, Immunofluorescence (IF), Immunofluorescence Staining and Detection, Ionic Homeostasis and Signaling, pH Detection

Order Info

Shipping Condition: Wet Ice

Specifications

Content And Storage Store in freezer (-5 to -30°C) and protect from light.
Detection Method Fluorescence
Product Type Growth Factor
Label or Dye Alexa Fluor Dyes
Product Line Alexa Fluor
Quantity 100 μg
Shipping Condition Wet Ice

For Research Use Only. Not for use in diagnostic procedures.