missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Epidermal Growth Factor, Biotin-XX Conjugate (biotin EGF)

Catalog No. E3477
Encompass
Change view
Click to view available options
Quantity:
20 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
E3477 20 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. E3477 Supplier Invitrogen™ Supplier No. E3477

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

This purified epidermal growth factor (EGF) contains a single biotin-XX molecule on the N-terminal amino acid

  • Biotin-XX EGF contains a long spacer arm that enhances the probe affinity for the EGF receptor and facilitates binding of a label such as a dye-streptavidin molecule, a Qdot™ nanocrystals-streptavidin molecule, or an enzyme-conjugated streptavidin
  • Labeled EGF has enabled scientists to investigate receptor-membrane interactions, study receptor distribution, calculate rate constants for the interaction of EGF with its receptor, and more
  • EGF molecule: 53 amino acids, MW=6,045 Da
  • Number of biotin-XX molecules on each EGF molecule: 1

Cell Analysis, Cell Viability, Proliferation and Function, Cellular Imaging, Endocytosis, Exocytosis and Phagocytosis, Immunofluorescence (IF), Immunofluorescence Staining and Detection, Nitro-Oxidative Stress, Reactive Oxygen Species

Order Info

Shipping Condition: Wet Ice

Specifications

Content And Storage Store in freezer (-5 to -30°C).
Detection Method Fluorescence
Form Solid
Product Type Growth Factor
Label or Dye Biotin & Other Haptens
Quantity 20 μg
Shipping Condition Wet Ice

For Research Use Only. Not for use in diagnostic procedures.