missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Click-IT™ O-GlcNAc Enzymatic Labeling System
GREENER_CHOICE

Catalog No. C33368
Encompass
Change view
Click to view available options
Quantity:
1 Kit
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C33368 1 Kit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. C33368 Supplier Invitrogen™ Supplier No. C33368

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

For in-vitro modification of O-GlcNAc modified proteins

The Click-iT O-GlcNAc Enzymatic Labeling System provides a highly sensitive and efficient method for the in-vitro modification of O-GlcNAc modified proteins. Proteins are enzymatically labeled utilizing the permissive mutant β-1,4-galactosyltransferase (Gal-T1 (Y289L)) which transfers azido-modified galactose (GalNAz) from UDP-GalNAz to O-GlcNAc residues. Then, via the chemoselective ligation or click reaction between an azide and an alkyne, the azido-labeled glycoproteins can then be detected with a Click-iT Glycoprotein Detection kit for gels (TAMRA or Dapoxyl™ alkyne) or Western blots (biotin alkyne). Labeling and detection can be completed in less than 24 hours and the Click-iT glycoprotein products are compatible with LC-MS/MS and Multiplexed Proteomics™ technologies for in-depth analyses of this important post-translational modification.

  • Less hazardous

Cell Analysis, Cell Structure, Click Chemistries, Glycoprotein Analysis with Click Chemistry, Labeling Chemistry, Mitochondria, Mitochondria Related Diseases, Mitochondrial Pathways, Protein and Antibody Labeling, Protein Gel Electrophoresis, Protein Gel Staining and Imaging, Proteins, Expression, Isolation and Analysis

Order Info

Shipping Condition: Wet ice

Specifications

Product Type O-GlcNAc Enzymatic Labeling System
Quantity 1 Kit
Content And Storage Store in refrigerator at 2°C to 8°C
Detection Method Biotin-based, Fluorescence
Shipping Condition Wet Ice
Product Line Click-iT
Label or Dye Alexa Fluor 488, Alexa Fluor 555, Alexa Fluor 594, Alexa Fluor 647, Biotin, Oregon Green 488, TMR (Tetramethylrhodamine)
Labeling Target Proteins

For Research Use Only. Not for use in diagnostic procedures.