missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Alexa Fluor™ 555-aha-dUTP, 1 mM in TE buffer

Catalog No. A32762
Encompass
Change view
Click to view available options
Quantity:
50 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A32762 50 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. A32762 Supplier Invitrogen™ Supplier No. A32762

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

For brighter conjugates

Labeled 5-aminohexylacrylamido-dUTP (aha-dUTP) nucleotides are modified at the C-5 position of uridine with a unique hexylacrylamide linker, which serves as a spacer between the nucleotide and the dye. This spacer reduces interactions between the nucleotide and the dye, resulting in brighter conjugates and increased hapten accessibility for secondary detection reagents. Labeled nucleotides can be used to generate labeled nucleic acid hybridization probes for many molecular biology and molecular cytogenetics applications, including multicolor techniques.

Array CGH, DNA and RNA Purification and Analysis, DNA Labeling, Gene Expression Analysis and Genotyping, Genotyping and Genomic Profiling, Nucleic Acid Labeling and Oligo Synthesis

Order Info

Shipping Condition: Wet Ice

Specifications

Product Type Labeled Nucleotide
Concentration 1 mM
Content And Storage Store in freezer (-5 to -30°C) and protect from light.
Label or Dye Alexa Fluor Dye
Modification AHA (5-aminohexylacrylamido)
Shipping Condition Wet Ice
Product Line Alexa Fluor
For Use With (Application) Molecular Biology
Quantity 50 μL
Can dU-containing DNA produced by amplification with dUTP be cut by restriction enzymes?

You will need to confirm with the manufacturer of the restriction enzyme you are using to find out whether or not it can recognize dUTP-containing DNA.

For Research Use Only. Not for use in diagnostic procedures.