missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Moclobemide

$522.29 - $1161.65

Chemical Identifiers

CAS 71320-77-9
Molecular Formula C13H17ClN2O2
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AC465800010
View Documents
Thermo Scientific Chemicals
465800010
1 g
Each for $1,161.65
Only null left
 
AC465802500
View Documents
Thermo Scientific Chemicals
465802500
250 mg
Each for $522.29
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

71320-77-9
C13H17ClN2O2

Specifications

71320-77-9
135.0°C to 138.0°C
268.74
ClC1=CC=C(C=C1)C(=O)NCCN1CCOCC1
268.74
1 g
C13H17ClN2O2
YHXISWVBGDMDLQ-UHFFFAOYSA-N
4-chloro-N-[2-(morpholin-4-yl)ethyl]benzamide
Moclobemide
Videos
Safety and Handling

Safety and Handling

GHS H Statement Harmful if swallowed.

Causes skin irritation.

Causes serious eye damage.

May cause respiratory irritation.

GHS P Statement Wear protective gloves/protective clothing/eye protection/face protection.

Immediately call a POISON CENTER or doctor/physician.

IF IN EYES: Rinse cautiously with water for several minutes.

Remove contact lenses, if present and easy to do.

Continue rinsing.

IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.

IF SWALLOWED: rinse mouth.

Do NOT induce vomiting.

IF ON SKIN: Wash with plenty of soap and water.

GHS Signal Word: Danger

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only