accessibility menu, dialog, popup

UserName

ClOHNNO

CAS RN 71320-77-9

IUPAC Name: 4-chloro-N-[2-(morpholin-4-yl)ethyl]benzamide
Synonyms: moclobemide betapharm brand of moclobemide bull brand of moclobemide bc brand of moclobemide terry white chemists brand of moclobemide novomoclobemide moclobemide kendrick brand moclamide moclamine arima
Molecular Weight (g/mol): 268.74
Molecular Formula: C13H17ClN2O2
InChi Key: YHXISWVBGDMDLQ-UHFFFAOYSA-N
SMILES: ClC1=CC=C(C=C1)C(=O)NCCN1CCOCC1
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results